Learn More
Thermo Scientific Chemicals Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 40 x 30 x 26;Vol (ml), 20
Product Code. 15423515
Change view
Click to view available options
Capacity (Metric):
6 mL
20 mL
40 mL
60 mL
110 mL
600 mL
Diameter (Metric) Outer:
20mm
30mm
35mm
40mm
50mm
85mm
Diameter (Metric) Inner:
16mm
26mm
30mm
36mm
44mm
73mm
Height (Metric):
30mm
40mm
50mm
60mm
75mm
150mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) unitSize
15423515 20 mL 30mm 26mm 40mm Each
15473515 600 mL 85mm 73mm 150mm Each
15453515 110 mL 50mm 44mm 75mm Each
15433515 40 mL 35mm 30mm 50mm Each
15443515 60 mL 40mm 36mm 60mm Each
15413515 6 mL 20mm 16mm 30mm Each
6 options
Product Code. 15423515
Brand: Thermo Scientific Chemicals 032952.KF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 26mm
Material Al-23
Quantity 1 Piece
Capacity (Metric) 20 mL
Diameter (Metric) Outer 30mm
Height (Metric) 40mm
Product Type Cylindrical Al-23 Crucibles
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.