Learn More
Cyhalofop-butyl, TRC
Specifications
Specifications
| Chemical Name or Material | Cyhalofop-butyl |
| CAS | 122008-85-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H20 F N O4 |
| Quantity | 1 g |
| Synonym | Propanoic acid, 2-[4-(4-cyano-2-fluorophenoxy)phenoxy]-, butyl ester, (R)-, Clincher, Clincher CA, Cyhalofop-butyl, XDE 537 |
| InChI Formula | InChI=1S/C20H20FNO4/c1-3-4-11-24-20(23)14(2)25-16-6-8-17(9-7-16)26-19-10-5-15(13-22)12-18(19)21/h5-10,12,14H,3-4,11H2,1-2H3/t14-/m1/s1 |
| SMILES | CCCCOC(=O)[C@@H](C)Oc1ccc(Oc2ccc(cc2F)C#N)cc1 |
| IUPAC Name | butyl (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoate |
| Molecular Weight (g/mol) | 357.38 |
| Show More |