missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYGB Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812911
This item is not returnable.
View return policy
Description
CYGB Monoclonal antibody specifically detects CYGB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CYGB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WWM9, Q921A4, Q9CX80 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 114757, 114886, 170520 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6G6 | |
| Unconjugated | |
| CYGB | |
| 3110001K20Rik; Cygb; Cytoglobin; HGb; Histoglobin; Staap; STAP; stellate cell activation associated protein; stellate cell activation-associated protein | |
| Full length human recombinant protein of human CYGB (NP_599030) produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction