missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetylene, 97%

Product Code. p-7035714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480248 5 g 5g
15490248 25 g 25g
15400258 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480248 Supplier Thermo Scientific Alfa Aesar Supplier No. H53440.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6746-94-7 | C5H6 | 66.103 g/mol

Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6746-94-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD02181090
InChI Key NPTDXPDGUHAFKC-UHFFFAOYSA-N
Synonym cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
PubChem CID 138823
IUPAC Name ethynylcyclopropane
SMILES C#CC1CC1

Specifications

Density 0.781
Boiling Point 51°C
Flash Point −17°C (−1°F)
Refractive Index 1.429
Quantity 5 g
UN Number UN3295
Solubility Information Slightly soluble in water.
Formula Weight 66.1
Percent Purity 97%
Chemical Name or Material Cyclopropylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.