missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylboronic acid, 95%

Product Code. 11416150
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11416150

Brand: Thermo Scientific Alfa Aesar H27081.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 63076-51-7 | C5H11BO2 | 113.951 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
Molecular Weight (g/mol) 113.951
MDL Number MFCD01074541
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O

Specifications

Quantity 1 g
Formula Weight 113.95
Percent Purity 95%
Chemical Name or Material Cyclopentylboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.