Learn More
Cyclopentanemethanol, 98%, Thermo Scientific™
Product Code. p-97359 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
Product Code. 10775021
Brand: Thermo Scientific Acros 147150050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO
Specifications
Color Colorless
Density 0.9200g/mL
Boiling Point 162°C to 163°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H9CH2OH
Refractive Index 1.4565 to 1.4585
Quantity 5 g
Beilstein 06, 9
Specific Gravity 0.92
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentanemethanol, 96%