missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienylrhenium tricarbonyl, 99%, Thermo Scientific™

Product Code. 11387768
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387768 2 g 2g
11377768 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387768 Supplier Thermo Scientific Alfa Aesar Supplier No. 033655.04
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12079-73-1
Molecular Formula C8H8O3Re
Molecular Weight (g/mol) 338.36
MDL Number MFCD00058885
InChI Key NEXLWSRPYZNSNH-UHFFFAOYSA-N
Synonym cyclopentadienylrhenium tricarbonyl, rhenium, tricarbonyl eta5-2,4-cyclopentadien-1-yl

Specifications

Color White
Quantity 2 g
UN Number UN3466
Solubility Information Insoluble in water.
Formula Weight 335.33
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Cyclopentadienylrhenium tricarbonyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.