Learn More
Cyclohexylurea, 98%
Product Code. 11471397
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471397 25 g 25g
11481397 100 g 100g
11491397 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11471397 Supplier Thermo Scientific Alfa Aesar Supplier No. B24915.14
Request Bulk or Custom Format
CAS: 698-90-8 | C7H14N2O | 142.202 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 698-90-8
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00014286
InChI Key WUESWDIHTKHGQA-UHFFFAOYSA-N
Synonym 1-cyclohexylurea, urea, cyclohexyl, n-cyclohexylurea, n-cyclohexlurea, cyclohexyl urea, amino-n-cyclohexylamide, cyclohexyl-urea, monocyclohexylurea, n'-cyclohexylurea, n'-cyclohexyl urea
PubChem CID 69684
IUPAC Name cyclohexylurea
SMILES C1CCC(CC1)NC(=O)N
Specifications
Melting Point 191°C to 195°C
Quantity 25 g
Beilstein 636914
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Cyclohexylurea