Learn More
Cyclohexyl p-toluenesulfonate, 97%
Product Code. 11364835
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11364835
Brand: Thermo Scientific Alfa Aesar L17952.14
Request Bulk or Custom Format
CAS: 953-91-3 | C13H18O3S | 254.344 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 953-91-3
Molecular Formula C13H18O3S
Molecular Weight (g/mol) 254.344
MDL Number MFCD00014291
InChI Key OHHPZPDQZMUTCA-UHFFFAOYSA-N
Synonym cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06
PubChem CID 13722
IUPAC Name cyclohexyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2
Specifications
Melting Point 45°C to 46°C
Quantity 25 g
Beilstein 2217391
Formula Weight 254.35
Percent Purity 97%
Chemical Name or Material Cyclohexyl p-toluenesulfonate