Learn More
Cyclohexanone, Supelco™
Product Code. 15877140
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
190 L
Unit Size:
1 L
10 L
190 L
2.5 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877140 190 L 190 L
15847140 1 L 1 L
15864310 2.5 L 2.5 L
15857140 10 L 10 L
15867140 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15877140 Supplier Supelco Supplier No. 1028889191
 Restricted Item
Estimated Shipment: 19-11-2026
EMPLURA™
EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-94-1
Specifications
Melting Point -31°C
Density 0.95 g/cm3 at 20°C
Quantity 190 L
MDL Number MFCD00001625
Synonym Cyclohexanone, Pimelic ketone, Cyclohexanone
Boiling Point 155°C/1013 hPa
Molecular Weight (g/mol) 98.14
Grade Research
Chemical Name or Material Cyclohexanone