Learn More
α-Cyclodextrine, Millipore™
Specifications
Specifications
| CAS | 10016-20-3 |
| Melting Point | 278°C |
| MDL Number | MFCD00078207 |
| Quantity | 50 g |
| Synonym | α-Cyclodextrine, Schardinger α-dextrin, Cyclohexaamylose, Cyclomaltohexaose, α-Cyclodextrine |
| Molecular Weight (g/mol) | 972.84 |
| Grade | Research |