Learn More
Cyclin E1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11554-1-AP-150UL
 View more versions of this product
Product Code. 16848713
  • 4836.10 NOK / 150µL
Description
Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are expressed only in tumors but not in the normal tissues, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.
Specifications
Specifications
Cyclin E1
Polyclonal
Unconjugated
CCNE1
CCNE, CCNE1, cyclin E1, G1/S specific cyclin E1
Rabbit
Antigen Affinity Chromatography
RUO
12447, 898
-20°C
Liquid
Flow Cytometry, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P24864, Q61457
CCNE1
Cyclin E1 Fusion Protein Ag2110
150 μL
Primary
Human, Mouse