missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11554-1-AP-20UL
This item is not returnable.
View return policy
Description
Cyclin E is a regulatory subunit of Cdk2 and controls G1 / S transition during the mammalian cell cycle. Multiple isoforms of Cyclin E are expressed only in tumors but not in the normal tissues, suggesting a post transcriptional regulation of Cyclin E. In vitro analyses indicated that these truncated variant isoforms of Cyclin E are able to phosphorylate histone H1. Alterations in Cyclin E protein have been implicated as indicators of worse prognosis in various cancers.Specifications
| Cyclin E1 | |
| Polyclonal | |
| Unconjugated | |
| CCNE1 | |
| CCNE, CCNE1, cyclin E1, G1/S specific cyclin E1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12447, 898 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P24864, Q61457 | |
| CCNE1 | |
| Cyclin E1 Fusion Protein Ag2110 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?