Learn More
CYB5R3 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10894-1-AP-20UL
This item is not returnable.
View return policy
Description
CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occurring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism and, in erythrocytes, methemoglobin reduction.Specifications
| CYB5R3 | |
| Polyclonal | |
| Unconjugated | |
| CYB5R3 | |
| B5R, CYB5R3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1, Diaphorase 1, NADH cytochrome b5 reductase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100524254, 109754, 1727, 25035 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00387, P20070, Q9DCN2 | |
| CYB5R3, LOC100524254 | |
| CYB5R3 Fusion Protein Ag1339 | |
| 20 μL | |
| Primary | |
| Human, Pig, Rat, Mouse | |
| Antibody | |
| IgG |