Learn More
Cyanuric acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Cyanuric acid |
| CAS | 108-80-5 |
| Melting Point | 320°C to 330°C (decomposition) |
| Density | 2.5 g/cm³ at 20°C |
| Quantity | 50 kg |
| Molecular Formula | C3N3(OH)3 |
| Synonym | Cyanuric acid, 1,3,5-Triazine-2,4,6-triol, Isocyanuric acid, Cyanuric acid |
| Molecular Weight (g/mol) | 129.07 |
| Grade | Not Available |