Learn More
Thermo Scientific™ Cy5 Dye Calibration Plate, 384-well
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 plate
Unit Size:
1 set
Description
- Used to maintain the QuantStudio Dx Real-Time PCR System with 384-well block
- Please see the instrument user guide for full instructions on use
Specifications
Specifications
| Block Format | 384-well |
| Content And Storage | Store at -20°C ± 5°C. |
| For Use With (Equipment) | QuantStudio™ Dx Real Time PCR Instrument |
| Product Type | Calibration Plate |
| For Use With (Application) | Pure Dye Calibration |
| Shipping Condition | Dry Ice |
| Quantity | 1 plate |
| Plate Compatibility | 384-well |
| Calibration Type | Pure Dye Calibration |
| Block Configurations | 384-well |