missing translation for 'onlineSavingsMsg'
Learn More

CY-09, MedChemExpress

Product Code. p-200100322
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476607

Brand: Medchem Express HY103666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CY-09 is a selective and direct NLRP3 inhibitor. CY-09 directly binds to the ATP-binding motif of NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, resulting in the suppression of NLRP3 inflammasome assembly and activation.

CY-09 is a selective and direct NLRP3 inhibitor. CY-09 directly binds to the ATP-binding motif of NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, resulting in the suppression of NLRP3 inflammasome assembly and activation.

Specifications

Chemical Name or Material CY-09
Physical Form Solid
CAS 1073612-91-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H12F3NO3S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 150 mg/mL (354.25 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(C1=CC=C(/C=C2SC(N(CC3=CC=CC(C(F)(F)F)=C3)C\2=O)=S)C=C1)=O
Molecular Weight (g/mol) 423.43
Formula Weight 423.43
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.