Learn More
CXCR4 Mouse anti-Human, Clone: 12G5, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB170-SP
Encompass
Product Code. 15764414
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CXCR4 Monoclonal specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-reported.