missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR1/IL-8RA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2525.00 NOK
Specifications
| Antigen | CXCR1/IL-8RA |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1 - 3 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CXCR1/IL-8RA Polyclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Western Blot.Specifications
| CXCR1/IL-8RA | |
| Western Blot 1 - 3 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P25024 | |
| 3577 | |
| This antibody was raised against a synthetic peptide (FLARHRVTSYTSC) corresponding to amino acids 330-341 of human CXCR1 protein. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| CXCR1 | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title