missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR1/IL-8RA Antibody (1122A) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8628JF669
4522.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Mouse samples. It is validated for Flow Cytometry
Specifications
| CXCR1/IL-8RA | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| Mouse CXCR1/IL-8 RA Peptide, Accession # Q810W6 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry | |
| 1122A | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 3577 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction