missing translation for 'onlineSavingsMsg'
Learn More

CXCL8/IL-8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16846854 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16846854 150 μL 150µL
16856854 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16846854

Brand: Proteintech 170381AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CXCL8/IL-8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL8/IL-8. This antibody recognizes human antigen. The CXCL8/IL-8 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Antigen CXCL8/IL-8
Applications Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Concentration 300 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 3576
Target Species Human
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.