Learn More
CXCL8/IL-8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17038-1-AP-20UL
 View more versions of this product
Product Code. 16856854
  • 1710.33 NOK / 20µL
Description
The CXCL8/IL-8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL8/IL-8. This antibody recognizes human antigen. The CXCL8/IL-8 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
CXCL8/IL-8
Polyclonal
Unconjugated
20 μL
3576
IgG
Immunohistochemistry, Immunofluorescence
300 μg/mL