Learn More
CXCL8/IL-8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17038-1-AP-20UL
Description
The CXCL8/IL-8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL8/IL-8. This antibody recognizes human antigen. The CXCL8/IL-8 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| CXCL8/IL-8 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 3576 | |
| IgG |
| Immunohistochemistry, Immunofluorescence | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human |