missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXADR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11777-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.Specifications
| CXADR | |
| Polyclonal | |
| Unconjugated | |
| CXADR | |
| CAR, CVB3 binding protein, CXADR, HCAR, HCVADR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13052, 1525, 89843 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P78310, P97792, Q9R066 | |
| CXADR | |
| CXADR Fusion Protein Ag2369 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction