Learn More
QIAGEN Cutting Mat
Product Code. 11323365
Change view
Click to view available options
Quantity:
1 Pk.
Size:
Large (15.2 x 20.3 cm)
Small (6.3 x 7.6 cm)
Unit Size:
Each
Product Code. Quantity Size unitSize
11323365 1 Pk. Large (15.2 x 20.3 cm) Each
15342805 1 Pk. Small (6.3 x 7.6 cm) Each
2 options
Product Code. 11323365
Brand: QIAGEN WB100020
For precise sample disc removal from FTA cards.
  • The UniCore Cutting Mat ensures clean sample cuts from FTA/FTA Elute Cards and extends the life of the cutting tip of punch.
  • The matt is are designed for repeated, extended use and can be cleaned with alcohol and is available in two formats.
Specifications
Color Green
For Use With (Equipment) Micro Punch
For Use With (Application) Clean sample cuts from FTA/FTA™ Elute cards
Product Type Cuttting Mat
Quantity 1 Pk.
Size Large (15.2 x 20.3 cm)