Learn More
IKA™ MF 10.1 Impact Grinding Head
Product Code. 10791941 Shop All IKA Products
Change view
Click to view available options
Speed:
24.5 m/sec.
Unit Size:
Each
Product Code. Speed unitSize
10791941 24.5 m/sec. Each
1 options
Product Code. 10791941
Brand: IKA™ 0002870900
Only null left
Add to basket
Add to basket
For crushing fibrous substances such as paper and vegetation, but also for plastics and material with a low volume weight.
Before being discharged, the ground material passes through a sieve. This sieve is interchangeable and available in different hole sizes (not incl. with delivery). The ground material can then be collected using an NS 29 standard ground vessel.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cutting-grinding Head
Dimensions (L x W x H) 300 x 320 x 560 mm
For Use With (Equipment) For Plastics and Material with a Low Volume Weight
Height (Metric) 560 mm
Material Stainless Steel
Speed 24.5 m/sec.
Weight (Metric) 2.2 kg
Width (English) 12.6 in.
Width (Metric) 320 mm