Learn More
Epredia™ Cut Microscope Slide
Product Code. 17224884
Change view
Click to view available options
Thickness: 1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17224884 Thickness: 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17224884 Supplier Epredia™ Supplier No. AA00000102E01MNZ20
  • ONLINE EXCLUSIVE PRICE
    30.10 NOK
    26.83 NOK / Pack of 50
    Save 3.27 NOK
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Premium slides manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to be used in routine histology and microbiology applications.
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
TRUSTED_SUSTAINABILITY
Specifications
Material Soda Lime Glass
Thickness (Metric) 1 mm
Certifications/Compliance ISO8037/I
Coating Uncoated
Writing Surface No
Dimensions (L x W) 76 x 26 mm