Learn More
Ambion™ Custom Silencer™ siRNA
Product Code. 15581144
Change view
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
Product Code. Quantity unitSize
15581144 20 nmol 20nmol
15541144 20 nmol 20nmol
15501521 20 nmol Each
15551144 40 nmol 40nmol
15511521 40 nmol Each
15521521 40 nmol Each
15561144 40 nmol 40nmol
15531521 20 nmol Each
15571144 20 nmol 20nmol
15541521 20 nmol Each
15551521 20 nmol Each
15591144 20 nmol 20nmol
15501154 100 nmol 100nmol
15521154 1 umol 1µmol
14 options
Product Code. 15581144
Brand: Ambion™ AM16211
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Custom Silencer™ siRNA
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Spezifikation
Content And Storage Room temperature
Description Custom siRNA, 20nmol, CY3 Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Custom siRNA
Purity CY3
Quantity 20 nmol