Learn More
Ambion™ Custom Silencer™ siRNA
Product Code. 15551521
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
Product Code. 15551521
Brand: Ambion™ AM16212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Custom Silencer™ siRNA
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Custom siRNA, 20nmol, FAM Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
For Use With (Application) RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
Product Type Custom siRNA
Purity FAM
Quantity 20 nmol