Learn More
Invitrogen™ Custom Silencer™ siRNA
Description
Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Specifications
| Content And Storage | Store at or below -20°C. Do not store in a frost-free freezer. Dried oligonucleotides are shipped at ambient temperature. |
| Description | Custom siRNA, 20 nmol |
| RNAi Type | siRNA |
| Sample Type | RNA |
| Product Line | Ambion™, Silencer™ Select |
| Product Type | siRNA |
| Quantity | 20 nmol |