Learn More
Ambion™ Custom Select siRNA
Click to view available options
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Unit Size:
20nmol
250nmol
40nmol
5nmol
Each
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Specifications
| Content And Storage | Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C |
| Description | Custom Select siRNA, 5nmol, For RNAi, Epigenetics and Non-Coding RNA research, RNAi, siRNA |
| RNAi Type | siRNA |
| Final Product Type | Custom select siRNA |
| For Use With (Application) | RNAi, epigenetics and non-coding RNA research, RNAi, siRNA |
| Product Line | Silencer, Ambion |
| Product Type | Custom Select siRNA |
| Quantity | 5 nmol |
| Sample Size | 5nmol |