Learn More
Ambion™ Custom Select siRNA
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Specifications
| Content And Storage | Room temperature |
| Description | Custom Select siRNA, 20nmol, HPLC Purity, ≥95% Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities |
| For Use With (Application) | Knocking down gene expression in order to study protein function in a wide range of cell types |
| Product Type | Custom Select siRNA |
| Purity | HPLC |
| Quantity | 20 nmol |