Learn More
Ambion™ Custom RNA, Stealth Seq
Product Code. 10052312
Change view
Click to view available options
Quantity:
1 μmol
1.7 μmol
2 nmol
20 nmol
500 nmol
80 nmol
Unit Size:
1.7µmol
1µmol
20nmol
80nmol
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10052312 1 μmol 1µmol
10397782 20 nmol 20nmol
10664953 1.7 μmol 1.7µmol
10636683 80 nmol 80nmol
10022713 500 nmol Each
10337843 2 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10052312 Supplier Ambion™ Supplier No. 10650004
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Custom RNA, Stealth Seq
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Description RNA Stealth SEQ, 1μmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA Stealth SEQ
Quantity 1 μmol