Learn More
Ambion™ Custom RNA, Stealth, HPLC
Product Code. 10164353
Change view
Click to view available options
Product Type:
RNA HPLC In-vivo
Stealth RNA HPLC
Quantity:
20 nmol
5 nmol
500 nmol
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10164353 Stealth RNA HPLC 20 nmol Each
10523054 RNA HPLC In-vivo 20 nmol Each
10328033 Stealth RNA HPLC 500 nmol Each
10667923 RNA HPLC In-vivo 500 nmol Each
10522504 Stealth RNA HPLC 5 nmol Each
10174283 RNA HPLC In-vivo 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10164353 Supplier Ambion™ Supplier No. 100001871
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Custom RNA, Stealth, HPLC
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Stealth RNA HPLC, 20nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type Stealth RNA HPLC
Purity HPLC
Quantity 20 nmol