Learn More
Ambion™ Custom mirVana™ miRNA Mimic
Product Code. 15541124
Change view
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
Product Code. Quantity unitSize
15541124 20 nmol 20nmol
15531124 5 nmol 5nmol
15551124 250 nmol 250nmol
3 options
Product Code. 15541124
Brand: Ambion™ 4464069
Custom mirVana™ miRNA Mimic
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Customer-defined mirVana™ miRNA mimic, 20nmol, miRNA mimics are small, Chemically Modified Double-Stranded RNAs that mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-Regulation of miRNA Activity
Product Type Customer-defined mirVana™ miRNA mimic
Quantity 20 nmol