Learn More
Ambion™ Custom mirVana™ miRNA Inhibitor
Product Code. 15501134
Click to view available options
Quantity:
250 nmol
5 nmol
Unit Size:
250nmol
Each
Product Code. 15501134
Brand: Ambion™ 4464089
Custom mirVana™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation. Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description mirVana Custom miRNA Inhibitor, 250nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type mirVana Custom miRNA Inhibitor
Quantity 250 nmol