Learn More
Thermo Scientific™ Value DNA Oligo , Desalted, Dry, 24 , 25 nmol
Product Code. 15470704
Change view
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470704 25 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15470704 Supplier Thermo Scientific™ Supplier No. A15612
 Restricted Item
Estimated Shipment: 02-12-2026
Custom DNA Oligo, 25N, Desalted, Dry
Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications
Description DNA Oligo, 25 nmole, 25N, Dry, Desalted, Cost-effective and Fastest
Product Type DNA Oligo
Purification Method Desalted
Quantity 25 nmol
Form Dry