Learn More
Hettich Lab Technology™ Cushion
Product Code. 10412792
Click to view available options
:
For use with: Carrier 4890, 450mL/750mL Tube (4447, 4234-A), 650mL/750mL Bottle (0554, 0512)
Product Code. 10412792
Brand: Hettich Lab Technology™ 4451
Hettich Lab Technology™ Cushion can be used to temper the effect of forces on centrifuge tubes/bottles and their contents.
  • Precisely cut for fit
  • Protects carrier and sample vessel
Order Info
dimForUseWithEquipment
Specifications
Type Cushion
For Use With (Equipment) Carrier 4890, 450mL/750mL Tube (4447, 4234-A), 650mL/750mL Bottle (0554, 0512)
Tube Size 97 mm dia. Bottle