Learn More
Fisherbrand™ Curved Base Radiation Gamma Shields
Product Code. 12357177 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
450 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12357177 450 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12357177
Estimated Shipment: 05-04-2027
Gamma radiation shields with fixed 45 degree angles and a curved base of various dimensions, are ideal for protection from Gamma radiation.
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Radiation shielding Cleaver Scientific for protection from Gamma radiation, shield, fixed, 45 degree angle, curved base, dimensions 450h x 300w x 150d mm
Specifications
Product Type Gamma Shield
Width (Metric) 300 mm
Height (Metric) 450 mm
Thickness (Metric) 350 mm