Learn More
Micronova™ CurtainCleaner™ Irradiated Slipcovers
BIOPHARMAGREENER_CHOICE
Product Code. p-7202984 Shop All Micronova Products
Change view
Click to view available options
Dimensions:
14.25 x 2.5 in.
30.5 x 5.1 cm (12 x 2 in.)
Material:
Smooth Knit Polyester, Urethane Foam
Unit Size:
12 pairs
Pack of 12
Product Code. Dimensions Material unitSize
11922465 30.5 x 5.1 cm (12 x 2 in.) Smooth Knit Polyester, Urethane Foam Pack of 12
12775705 14.25 x 2.5 in. Smooth Knit Polyester, Urethane Foam 12 pairs
2 options
Product Code. 11922465
Brand: Micronova™ CCSC6IR
Only null left
Add to basket
Add to basket
These irradiated, 100% knit polyester slipcovers work with the Curtain Cleaner adapter to provide a simple method to sanitize cleanroom strip curtains by clamping the top of the strip and sliding down, effectively cleaning both sides at once.
The Micronova Irradiated CurtainCleaner™ SlipCovers work with the CurtainCleaner Adapter/Handle (CCH-48) to provide a simple and effective method to clean strip curtains in the cleanroom, without the need for a ladder or stool. The covers slide onto the dual 12-inch bars that clamp onto the curtain at the top and slide down to apply or 'squeegee' the cleaning agent on both sides of the curtain at once. Covers are available in smooth 100% knit polyester, with two thickness options. Gamma irradiated covers are ideal for aseptic areas.
  • Slipcovers are 100% knit polyester over thick foam
  • Cleans both sides of strip curtains at once
  • Excellent for cleaning and sanitizing
  • Covers fit over two 12-inch bar clamps
  • Gamma irradiated for aseptic areas
  • Launderable for re-use
  • Autoclavable
Order Info
Handle sold separately (Cat. No. 10-041-766A).
Specifications
Dimensions 30.5 x 5.1 cm (12 x 2 in.)
Product Type Irradiated Mop Slipcover
Color White
Length (English) 12 in.
Sterility Sterile
Length (Metric) 30.5 cm
Autoclavable Autoclavable
Width (Metric) 5.1 cm
Width (English) 2 in.
Resistant To Chemicals
Disposable Reusable
Care Instructions Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment) Micronova Curtain Cleaner Curtain Cleaning System
For Use With (Application) Cleaning and Sanitization Cleanroom Strip Curtains
Color-coded No
Material Smooth Knit Polyester, Urethane Foam