Learn More
Curcumin, Sigma-Aldrich™
Product Code. 15890901
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15890901 10 g 10g
15806640 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15890901 Supplier Sigma-Aldrich Supplier No. 8203540010
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 458-37-7
Specifications
Melting Point 170°C to 175°C
Molecular Formula [4-(OH)-3-(CH3O)C6H3CH=CHCO]2CH2
MDL Number MFCD00008365
Quantity 10 g
Synonym Curcumin, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Turmeric yellow, Diferuloylmethane, Curcumin
Molecular Weight (g/mol) 368.38
Grade Research