Learn More
R&D Systems™ Cultrex 3D Culture Matrix Rat Collagen I
Description
Functional Assays: Promotes cell attachment and spreading of HT-1080 human fibrosarcoma cells. Collagen I promotes attachment and growth of murine endothelial SVEC4-10 cells in 3-D culture. Gelling Assay: Type I collagen forms a firm gel at neutral pH and 37°C when diluted to 0.4 mg/ml.
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
Specifications
Specifications
| Product Type | Collagen I insoluble extracellular glycoproteins |
| Product Line | R&D Systems |
| Quantity | 100 mg |
| Endotoxin Level | <20.0EU per 1μg of the protein by the LAL method |
| Regulatory Status | RUO |
| Source | Rat tail tendons |
| Content And Storage | Product is stable for a minimum of 3 months if stored at 4 °C. Do Not Freeze. Storage Buffer: 20 mM Acetic Acid Sterility Testing: No bacterial or fungal growth detected after incubation at 37 °C for 14 days following USP XXIV Chapter 71 sterility test. No mycoplasma contamination detected by PCR. |
For Research Use Only. Not For Use In Diagnostic Procedures
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?