Learn More
Thermo Scientific™ Culti-Loops™ Streptococcus pneumoniae ATCC ™6303™
Product Code. 11983292
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Pack of 5
Product Code. 11983292
Brand: Thermo Scientific™ R4608221
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 6303TM*
Description Streptococcus pneumoniae ATCC™ 6303™
Format Culti-Loops
Product Type Culti-Loop
Target Organism Class Cocci
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations WDCM00005, [CIP 104225]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.