Learn More
Thermo Scientific™ Culti-Loops™ Staphylococcus simulans ATCC™ 27851™
Product Code. 12637659
Change view
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Each
Product Code. Quantity unitSize
12637659 5 Loops/Pack Each
1 options
Product Code. 12637659
Brand: Thermo Scientific™ R4607013
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 27851TM*
Description Staphylococcus simulans ATCC™ 27851™
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Cocci
Test Type Qualitative QC
Quantity 5 Loops/Pack
Strain Designations MK 148
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.