Learn More
Thermo Scientific™ Culti-Loops Salmonella Tranoroa Nctc 10252
Product Code. 12979068
Change view
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12979068 5 Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
If you would like to order, a signed ATCC agreement is required.
Product Code. 12979068 Supplier Thermo Scientific™ Supplier No. R4606010
If you would like to order, a signed ATCC agreement is required.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Culti-Loops are manufactured using proprietary gel preservation offering unique advantages over lyophilized organisms including: faster recovery, enhanced viability, and direct-streak-to-plate capabilities. More efficient recovery of organisms saves on laboratory resources and time. To recover the organism, simply moisten the gel film by placing the loop directly on warm agar for 10 to 15 seconds
Thermo Scientific™ Culti-Loops™ are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix. Convenience Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration. Performance Obtain enhanced viability and recovery with proprietary gel-preserved organisms. Compliance Over 400 Culti-Loops are Certified Reference Materials (CRM) manufactured by an Accredited Reference Material Producer Value Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms. Confidence Reach complete traceability with ATCC™ License Derivative strains. Efficiency Achieve accurate and convenient documentation with peel-off, transferable labels. Safety Reduce the risk of contamination and infection with zero organism handling.
TRUSTED_SUSTAINABILITY
Specifications
Manufacturing Origin United States
Sterility Non-sterile
Target Organism Class Bacteria
Test Type Qualitative QC
Quantity 5 Pk.