Learn More
Thermo Scientific™ Culti-Loops Salmonella Tranoroa Nctc 10252
Description
Culti-Loops are manufactured using proprietary gel preservation offering unique advantages over lyophilized organisms including: faster recovery, enhanced viability, and direct-streak-to-plate capabilities. More efficient recovery of organisms saves on laboratory resources and time. To recover the organism, simply moisten the gel film by placing the loop directly on warm agar for 10 to 15 seconds
Thermo Scientific™ Culti-Loops™ are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix. Convenience Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration. Performance Obtain enhanced viability and recovery with proprietary gel-preserved organisms. Compliance Over 400 Culti-Loops are Certified Reference Materials (CRM) manufactured by an Accredited Reference Material Producer Value Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms. Confidence Reach complete traceability with ATCC™ License Derivative strains. Efficiency Achieve accurate and convenient documentation with peel-off, transferable labels. Safety Reduce the risk of contamination and infection with zero organism handling.
Specifications
Specifications
| Manufacturing Origin | United States |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Qualitative QC |
| Quantity | 5 Pk. |