Learn More
Thermo Scientific™ Culti-Loops™ Paenibacillus polymyxa ATCC™ 43865™
Product Code. 15299467
Click to view available options
Quantity:
5 Loops/Pk.
Unit Size:
Pack of 5
Product Code. 15299467
Brand: Thermo Scientific™ R4601234
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 43865TM*
Certifications/Compliance Culti-Loops are CE Marked.
Description API 50 CHB. P. polymyxa ATCC™ 43865™
Detects Paenibacillus polymyxa
Product Type Single Organism
Target Organism Class Gram-positive Rods
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Strain Designations LRA 88.01.80
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.