Learn More
CUL2 Mouse anti-Human, Unconjugated, Clone: 1A6G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67175-1-IG-20UL
This item is not returnable.
View return policy
Description
The CUL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL2. This antibody recognizes human, rat antigen. The CUL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CUL2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A6G9 | |
| Unconjugated | |
| 20 μL | |
| 8453 | |
| IgG1 |