Learn More
Sartorius Cubis™ II High-Capacity Lab Balance
BIOPHARMA
Product Code. 16437045 Shop All Sartorius Products
Click to view available options
Capacity:
20,200 g
36,200 g
70,200 g
Includes:
Draft Shield O
Draft Shield O, QP1
Draft Shield O, QP99
Model:
CEU Verified balance with EC Type Approval Certificate (for EU except France)
S00 Standard version non-verified
Unit Size:
1 unit
Each
Product Code. 16437045
Brand: Sartorius MCA70201S2CEU0
The Cubis™ II High-Capacity Lab Balance with a maximum load between 11.2 kg and 70.2 kg and readability between 0.1 g and 1 g provides the ideal model for every application.
The New Generation of Modular Premium Laboratory Balances
  • Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
  • One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
  • All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
  • These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
  • It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Balance with Type Approval Certificate for China
Connection Type D-sub connector, 9-pin
Capacity 70,200 g
Display Type Color Touchscreen
Readability 100 mg
Includes Draft Shield O
Material Aluminum, plastic PBT, Optiwhite float glass and stainless steel
Model CEU Verified balance with EC Type Approval Certificate (for EU except France)
Stabilization Time 1.5 sec.
Type Electronic Toploading Balance
Electrical Requirements 100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A