Learn More
Sartorius Cubis™ II Analytical Lab Balance
BIOPHARMA
Product Code. 16425005 Shop All Sartorius Products
Click to view available options
Capacity:
120 g
120 g, 240 g, 520 g
220 g
320 g
80 g, 160 g, 320 g
Model:
Balance with Draft Shield A
Balance with Draft Shield I
Balance with Draft Shield U
Readability:
0.1 g, 0.2 g, 0.5 g
0.1 mg
0.1 mg, 0.2 mg, 0.5 mg
Unit Size:
Each
Product Code. 16425005
Brand: Sartorius MCA324P2S00A QP99
Maximum weighing capacity between 120 g and 520 g in combination with a readability of 0.1 mg.
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Balance with Type Approval Certificate for China
Connection Type D-sub connector, 9-pin
Capacity 80 g, 160 g, 320 g
Display Type Color Touchscreen
Readability 0.1 mg, 0.2 mg, 0.5 mg
Includes Internal Calibration
Material Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel
Model Balance with Draft Shield A
Stabilization Time 1 sec.
Type Analytical Balance
Electrical Requirements 100/240 V (± 10%), 50/60 Hz, 1.0 A