missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTTNBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17893-1-AP-150UL
This item is not returnable.
View return policy
Description
CTTB2 is a protein with six ankyrin repeats and several proline-rich regions. A similar gene in rat interacts with a central regulator of the actin cytoskeleton.Specifications
| CTTNBP2 | |
| Polyclonal | |
| Unconjugated | |
| CTTNBP2 | |
| C7orf8, cortactin binding protein 2, CORTBP2, CTTNBP2, KIAA1758, Orf4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30785, 83992 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B9EJA2, Q8WZ74 | |
| Cttnbp2 | |
| CTTNBP2 Fusion Protein Ag12219 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction