missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTSW (Human) Recombinant Protein
Human CTSW full-length ORF ( AAH48255, 22 a.a. - 376 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001521-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: IRGPLRAQDLGPQPLELKEAFKLFQIQFNRSYLSPEEHAHRLDIFAHNLAQAQRLQEEDLGTAEFGVTPFSDLTEEEFGQLYGYRRAAGGVPSMGREIRSEEPEESVPFSCDWRKVAGAISPIKDQKNCNCCWAMAAAGNIETLWRISFWDFVDVSVQELLDCGRCGDGCHGGFVWDAFITVLNNSGLASEKDYPFQGKVRAHRCHPKKYQKVAWIQDFIMLQNNEHRIAQYLATYGPITVTINMKPLQLYRKGVIKATPTTCDPQLVDHSVLLVGFGSVKSEEGIWAETVSSQSQPQPPHPTPYWILKNSWGAQWGEKGYFRLHRGSNTCGITKFPLTARVQKPDMKPRVSCPP
Specifications
AAH48255 | |
cathepsin W | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTSW | |
GST |
1521 | |
Wheat germ expression system | |
25 μg | |
LYPN | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |